COM448684996 Combi-Blocks, Inc.

4-(3-Nitrophenyl)phenol

  • MDL number MFCD09909462
  • Cas Number : 53059-30-6
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1635
Availability details for
Quantity
  • Availability
  • Add to Cart