COM448685003 Combi-Blocks, Inc.

3-(4-Fluoro-3-methoxyphenyl)phenol

  • MDL number MFCD18312866
  • Cas Number : 1261957-92-9
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1658

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1658
Availability details for
Quantity
  • Availability
  • Add to Cart