COM448685005 Combi-Blocks, Inc.

4-(3-Chloro-4-methylphenyl)phenol

  • MDL number MFCD00509341
  • Cas Number : 191724-14-8
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1671
Availability details for
Quantity
  • Availability
  • Add to Cart