COM448685006 Combi-Blocks, Inc.

2-(Naphthalen-2-yl)phenol

  • MDL number MFCD18312884
  • Cas Number : 78210-35-2
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1681
Availability details for
Quantity
  • Availability
  • Add to Cart