COM448685010 Combi-Blocks, Inc.

4-(3-Chloro-4-fluorophenyl)phenol

  • MDL number MFCD11931984
  • Cas Number : 22510-31-2
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1695
Availability details for
Quantity
  • Availability
  • Add to Cart