COM448685012 Combi-Blocks, Inc.

3-[Benzo(b)thiophen-2-yl]phenol

  • MDL number MFCD18312902
  • Cas Number : 95935-60-7
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1700
Availability details for
Quantity
  • Availability
  • Add to Cart