COM448685013 Combi-Blocks, Inc.

4-[Benzo(b)thiophen-2-yl]phenol

  • MDL number MFCD18312903
  • Cas Number : 65540-08-1
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1701
Availability details for
Quantity
  • Availability
  • Add to Cart