COM448685014 Combi-Blocks, Inc.

3-(4-t-butylphenyl)phenol

  • MDL number MFCD06802328
  • Cas Number : 893737-20-7
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1703

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1703
Availability details for
Quantity
  • Availability
  • Add to Cart