COM448685015 Combi-Blocks, Inc.

4-(4-T-butylphenyl)phenol

  • MDL number MFCD06802300
  • Cas Number : 19812-92-1
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1704

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1704
Availability details for
Quantity
  • Availability
  • Add to Cart