COM448685027 Combi-Blocks, Inc.

2-(4-Carboxy-3-fluorophenyl)phenol

  • MDL number MFCD18312929
  • Cas Number : 1261975-31-8
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1741
Availability details for
Quantity
  • Availability
  • Add to Cart