COM448685030 Combi-Blocks, Inc.

4-(5-Carboxy-2-fluorophenyl)phenol

  • MDL number MFCD11933453
  • Cas Number : 1181608-84-3
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1746
Availability details for
Quantity
  • Availability
  • Add to Cart