COM448685031 Combi-Blocks, Inc.

2-(Naphthalen-1-yl)phenol

  • MDL number MFCD18312933
  • Cas Number : 101277-90-1
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1747
Availability details for
Quantity
  • Availability
  • Add to Cart