COM448685034 Combi-Blocks, Inc.

2-(3-Trifluoromethylphenyl)phenol

  • MDL number MFCD06802853
  • Cas Number : 893738-60-8
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1768
Availability details for
Quantity
  • Availability
  • Add to Cart