COM448685036 Combi-Blocks, Inc.

4-(3-Trifluoromethylphenyl)phenol

  • MDL number MFCD06801898
  • Cas Number : 191724-12-6
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1770
Availability details for
Quantity
  • Availability
  • Add to Cart