COM448685039 Combi-Blocks, Inc.

4-(4-Trifluoromethylphenyl)phenol

  • MDL number MFCD06802299
  • Cas Number : 10355-13-2
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1773

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1773
Availability details for
Quantity
  • Availability
  • Add to Cart