COM448685046 Combi-Blocks, Inc.

4-(3,5-Dichlorophenyl)phenol

  • MDL number MFCD11932332
  • Cas Number : 4291-32-1
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1788
Availability details for
Quantity
  • Availability
  • Add to Cart