COM448685048 Combi-Blocks, Inc.

4-[3-(N,N-Dimethylaminocarbonyl)phenyl]phenol

  • MDL number MFCD18312956
  • Cas Number : 1261896-44-9
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1791

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1791
Availability details for
Quantity
  • Availability
  • Add to Cart