COM448685056 Combi-Blocks, Inc.

3-(4-Methylsulfonylphenyl)phenol

  • MDL number MFCD06802344
  • Cas Number : 893737-42-3
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1823

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1823
Availability details for
Quantity
  • Availability
  • Add to Cart