COM448685067 Combi-Blocks, Inc.

4-[4-(Ethylcarbamoyl)-3-fluorophenyl]phenol

  • MDL number MFCD18313009
  • Cas Number : 1261894-26-1
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1851
Availability details for
Quantity
  • Availability
  • Add to Cart