COM448685068 Combi-Blocks, Inc.

2-[4-(Ethoxycarbonyl)-3-fluorophenyl]phenol

  • MDL number MFCD18313010
  • Cas Number : 1261975-80-7
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1852
Availability details for
Quantity
  • Availability
  • Add to Cart