COM448685069 Combi-Blocks, Inc.

2-(3-Pyrrolidinocarbonylphenyl)phenol

  • MDL number MFCD18313025
  • Cas Number : 1261965-79-0
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1867

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1867
Availability details for
Quantity
  • Availability
  • Add to Cart