COM448685073 Combi-Blocks, Inc.

4-(3-Benzyloxyphenyl)phenol

  • MDL number MFCD06802269
  • Cas Number : 889950-85-0
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1881
Availability details for
Quantity
  • Availability
  • Add to Cart