COM448685078 Combi-Blocks, Inc.

4-(3-N,N-Dimethylsulfamoylphenyl)phenol

  • MDL number MFCD18313039
  • Cas Number : 1261947-72-1
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1890
Availability details for
Quantity
  • Availability
  • Add to Cart