COM448685079 Combi-Blocks, Inc.

4-(4-N,N-Dimethylsulfamoylphenyl)phenol

  • MDL number MFCD18313042
  • Cas Number : 1261900-34-8
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1893
Availability details for
Quantity
  • Availability
  • Add to Cart