COM448685082 Combi-Blocks, Inc.

4-(3-BOC-Aminophenyl)phenol

  • MDL number MFCD18313048
  • Cas Number : 1261958-25-1
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1899
Availability details for
Quantity
  • Availability
  • Add to Cart