COM448685087 Combi-Blocks, Inc.

2-Methyl-5-phenylphenol

  • MDL number MFCD03265274
  • Cas Number : 105902-32-7
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1922

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1922
Availability details for
Quantity
  • Availability
  • Add to Cart