COM448685088 Combi-Blocks, Inc.

4-(Furan-2-yl)-2-methylphenol

  • MDL number MFCD17677675
  • Cas Number : 1255638-84-6
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1924

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1924
Availability details for
Quantity
  • Availability
  • Add to Cart