COM448685091 Combi-Blocks, Inc.

5-(3-Cyanophenyl)-3-methylphenol

  • MDL number MFCD18313106
  • Cas Number : 1261896-62-1
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1968
Availability details for
Quantity
  • Availability
  • Add to Cart