COM448685092 Combi-Blocks, Inc.

4-(4-Cyanophenyl)-2-methylphenol

  • MDL number MFCD17677586
  • Cas Number : 460748-08-7
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1969

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1969
Availability details for
Quantity
  • Availability
  • Add to Cart