COM448685093 Combi-Blocks, Inc.

4-(3-Formylphenyl)-2-methylphenol

  • MDL number MFCD18313112
  • Cas Number : 1261948-09-7
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1975
Availability details for
Quantity
  • Availability
  • Add to Cart