COM448685094 Combi-Blocks, Inc.

4-(4-Methoxyphenyl)-2-methylphenol

  • MDL number MFCD17677743
  • Cas Number : 103594-29-2
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2011
Availability details for
Quantity
  • Availability
  • Add to Cart