COM448685096 Combi-Blocks, Inc.

4-(4-Chlorophenyl)-2-methylphenol

  • MDL number MFCD18313188
  • Cas Number : 1261916-91-9
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2056
Availability details for
Quantity
  • Availability
  • Add to Cart