COM448685098 Combi-Blocks, Inc.

4-(3-Carboxyphenyl)-2-methylphenol

  • MDL number MFCD18313223
  • Cas Number : 1261976-44-6
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2092
Availability details for
Quantity
  • Availability
  • Add to Cart