COM448685099 Combi-Blocks, Inc.

4-(4-Methoxy-3-methylphenyl)-2-methylphenol

  • MDL number MFCD17677783
  • Cas Number : 1255636-23-7
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2107
Availability details for
Quantity
  • Availability
  • Add to Cart