COM448685107 Combi-Blocks, Inc.

3-Fluoro-5-(furan-2-yl)phenol

  • MDL number MFCD18313531
  • Cas Number : 1261996-61-5
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2408

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2408
Availability details for
Quantity
  • Availability
  • Add to Cart