COM448685113 Combi-Blocks, Inc.

4-(4-Fluorophenyl)-3-fluorophenol

  • MDL number MFCD14699562
  • Cas Number : 349665-92-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2455
Availability details for
Quantity
  • Availability
  • Add to Cart