COM448685121 Combi-Blocks, Inc.

5-(2-Carboxythiophene-4-yl)-3-fluorophenol

  • MDL number MFCD18313817
  • Cas Number : 1261936-09-7
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-2728
Availability details for
Quantity
  • Availability
  • Add to Cart