COM448685133 Combi-Blocks, Inc.

2-Cyano-4-(4-hydroxyphenyl)phenol

  • MDL number MFCD18314157
  • Cas Number : 1261973-27-6
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3071
Availability details for
Quantity
  • Availability
  • Add to Cart