COM448685147 Combi-Blocks, Inc.

2-Formyl-6-(2-trifluoromethylphenyl)phenol

  • MDL number MFCD18314811
  • Cas Number : 1261930-44-2
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3847
Availability details for
Quantity
  • Availability
  • Add to Cart