COM448685151 Combi-Blocks, Inc.

6-(2-Fluoro-5-methoxycarbonylphenyl)-2-formylphenol

  • MDL number MFCD18314838
  • Cas Number : 1261919-29-2
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-3886
Availability details for
Quantity
  • Availability
  • Add to Cart