COM448685156 Combi-Blocks, Inc.

4-(4-Carboxyphenyl)-2-methoxyphenol

  • MDL number MFCD18315107
  • Cas Number : 189161-82-8
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4175
Availability details for
Quantity
  • Availability
  • Add to Cart