COM448685158 Combi-Blocks, Inc.

4-(4-Carboxy-3-fluorophenyl)-2-methoxyphenol

  • MDL number MFCD18315233
  • Cas Number : 1261990-34-4
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4301
Availability details for
Quantity
  • Availability
  • Add to Cart