COM448685161 Combi-Blocks, Inc.

2-Chloro-5-(2-fluorophenyl)phenol

  • MDL number MFCD14701291
  • Cas Number : 1214379-85-7
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4515
Availability details for
Quantity
  • Availability
  • Add to Cart