COM448685163 Combi-Blocks, Inc.

2-Chloro-4-(4-fluorophenyl)phenol

  • MDL number MFCD11933401
  • Cas Number : 860616-69-9
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4520
Availability details for
Quantity
  • Availability
  • Add to Cart