COM448685166 Combi-Blocks, Inc.

3-Chloro-5-(2,4-difluorophenyl)phenol

  • MDL number MFCD18315532
  • Cas Number : 1262002-54-9
  • Molecular Weight : 240.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4621
Availability details for
Quantity
  • Availability
  • Add to Cart