COM448685168 Combi-Blocks, Inc.

3-Chloro-5-(3,5-difluorophenyl)phenol

  • MDL number MFCD18315536
  • Cas Number : 1261919-72-5
  • Molecular Weight : 240.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4627

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4627
Availability details for
Quantity
  • Availability
  • Add to Cart