COM448685173 Combi-Blocks, Inc.

3-Chloro-5-(2-fluoro-3-methoxyphenyl)phenol

  • MDL number MFCD18315593
  • Cas Number : 1261920-31-3
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4687
Availability details for
Quantity
  • Availability
  • Add to Cart