COM448685174 Combi-Blocks, Inc.

2-Chloro-4-(2-fluoro-4-methoxyphenyl)phenol

  • MDL number MFCD18315594
  • Cas Number : 1261931-63-8
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4688

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4688
Availability details for
Quantity
  • Availability
  • Add to Cart