COM448685177 Combi-Blocks, Inc.

5-(2-Carboxythiophene-4-yl)-3-chlorophenol

  • MDL number MFCD18315631
  • Cas Number : 1262003-03-1
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4726
Availability details for
Quantity
  • Availability
  • Add to Cart