COM448685179 Combi-Blocks, Inc.

2-Chloro-5-(4-methoxycarbonylphenyl)phenol

  • MDL number MFCD18315659
  • Cas Number : 1198422-80-8
  • Molecular Weight : 262.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-4755
Availability details for
Quantity
  • Availability
  • Add to Cart